Please login to your online account to display your discounted pricing

Pyrrolidine-1-carbonitrile, 97%, Maybridge

$88.65 - $172.48

Chemical Identifiers

Chemical Name or Material Pyrrolidine-1-carbonitrile
CAS 1530-88-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H8N2
Formula Weight 96.13
MDL Number MFCD00003165
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB00582EA

  SDS
fisher scientific company
SB00582EA
Amber Glass Bottle 10g Each for $88.65

SB00582EB

  SDS
fisher scientific company
SB00582EB
Amber Glass Bottle 25g Each for $172.48
Description & Specifications

Specifications

Formula Weight 96.13
Quantity 10g
Assay 97%
Density 0.9540g/mL
CAS Min % 97.0
Chemical Name or Material Pyrrolidine-1-carbonitrile
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C5H8N2
MDL Number MFCD00003165
CAS 1530-88-7
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.