missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyrrolidine-1-carbonitrile, 97%, Maybridge

Manufacturer:  Fisher Scientific Company SB00582EB

 View more versions of this product

Catalog No. SB00582EB


 

Specifications Description & Specifications

Specifications

96.13
97%
97.0
Pyrrolidine-1-carbonitrile
C5H8N2
Amber Glass Bottle
25g
0.9540g/mL
1530-88-7
MFCD00003165
97%

Chemical Identifiers

1530-88-7
97%
96.13
Pyrrolidine-1-carbonitrile
97.0
C5H8N2
MFCD00003165
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning