Learn More
Pyrrole, 99%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 157711000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Pyrrole | |
-23.0°C | |
0.9600g/mL | |
36°C | |
98.5% min. (GC) | |
C4H5N | |
MFCD00005216 | |
20,159 | |
15,8128 | |
Solubility in water: 60g/L (20°C). Other solubilities: insoluble in dilute alkalies, soluble in alcohol, ether and dilute acids, soluble in most organic solvents | |
N1C=CC=C1 | |
67.09 | |
1.317 mPa.s (20°C) | |
67.09 | |
Extra Pure |
109-97-7 | |
Brown to Colorless | |
129.0°C to 131.0°C | |
Authentic | |
Glass bottle | |
1.5080 to 1.5100 | |
100 g | |
0.96 | |
pyrrole, divinylenimine, azole, imidole, pyrrol, monopyrrole, divinyleneimine, 1-aza-2,4-cyclopentadiene, polypyrrole, unii-86s1zd6l2c | |
KAESVJOAVNADME-UHFFFAOYSA-N | |
1H-pyrrole | |
8027 | |
CHEBI:19203 | |
99% | |
Liquid |
Chemical Identifiers
109-97-7 | |
67.09 | |
KAESVJOAVNADME-UHFFFAOYSA-N | |
8027 | |
1H-pyrrole |
C4H5N | |
MFCD00005216 | |
pyrrole, divinylenimine, azole, imidole, pyrrol, monopyrrole, divinyleneimine, 1-aza-2,4-cyclopentadiene, polypyrrole, unii-86s1zd6l2c | |
CHEBI:19203 | |
N1C=CC=C1 |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Causes serious eye damage.
Toxic if swallowed.
Flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
GHS Signal Word: Danger
EINECSNumber : 203-724-7
RTECSNumber : UX9275000
TSCA : TSCA
Recommended Storage : Darkens on storage
RUO – Research Use Only