missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyrrole-3-carboxylic Acid, TCI America™

Manufacturer:  TCI America P18995G

 View more versions of this product

Catalog No. P18995G



Specifications

Specifications

Pyrrole-3-carboxylic Acid
148°C
931-03-3
MFCD00800594
DOYOPBSXEIZLRE-UHFFFAOYSA-N
1H-pyrrole-3-carboxylic acid
101030
111.10
≥95.0% (GC,T)
Undesignated
5g
C5H5NO2
pyrrole-3-carboxylic acid, pyrrole-3-carboxylicacid, 3-carboxy-1h-pyrrole, 1h-pyrrole-3-carboxylicacid, pubchem8345, 3-carboxypyrrole, 3-pyrrolecarboxylic acid, acmc-1agqb, 3-pyrrole-carboxylic acid, pyrrole 3-carboxylic acid
C1=CNC=C1C(=O)O
111.1
CHEBI:68076
Crystal-Powder at 20°C

Chemical Identifiers

931-03-3
111.1
DOYOPBSXEIZLRE-UHFFFAOYSA-N
101030
1H-pyrrole-3-carboxylic acid
C5H5NO2
MFCD00800594
pyrrole-3-carboxylic acid, pyrrole-3-carboxylicacid, 3-carboxy-1h-pyrrole, 1h-pyrrole-3-carboxylicacid, pubchem8345, 3-carboxypyrrole, 3-pyrrolecarboxylic acid, acmc-1agqb, 3-pyrrole-carboxylic acid, pyrrole 3-carboxylic acid
CHEBI:68076
C1=CNC=C1C(=O)O
Safety and Handling

Safety and Handling

TSCA : No

Documents