missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyrrole-3-carboxylic acid, 98+%

Manufacturer:  Alfa Aesar™ H55383MD

 View more versions of this product

Catalog No. AAH55383MD



Description

Description

Applications
Pyrrole-3-carboxylic acid is useful for the synthesis of cholecystokinin antagonists, benzopyran antihypertensives and azepinediones. It is utilized for the preparation of a poly(pyrrole-3-carboxylic acid) modified pencil graphite electrode and its usage in the determination of serotonin (SER) in blood serum and urine samples.

Solubility
Soluble in water.

Notes
Incompatible with strong bases, strong oxidizing agents, strong reducing agents.
Specifications

Specifications

Pyrrole-3-carboxylic acid
931-03-3
MFCD00800594
1h-pyrrole-3-carboxylicacid
DOYOPBSXEIZLRE-UHFFFAOYSA-N
1H-pyrrole-3-carboxylic acid
101030
111.1
250mg
C5H5NO2
108642
Soluble in water.
C1=CNC=C1C(=O)O
111.1
CHEBI:68076
≥98%

Chemical Identifiers

931-03-3
111.1
DOYOPBSXEIZLRE-UHFFFAOYSA-N
101030
1H-pyrrole-3-carboxylic acid
C5H5NO2
MFCD00800594
1h-pyrrole-3-carboxylicacid
CHEBI:68076
C1=CNC=C1C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents