Learn More
Pyrrole-3-carboxylic acid, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5538303
Description
Pyrrole-3-carboxylic acid is useful for the synthesis of cholecystokinin antagonists, benzopyran antihypertensives and azepinediones. It is utilized for the preparation of a poly(pyrrole-3-carboxylic acid) modified pencil graphite electrode and its usage in the determination of serotonin (SER) in blood serum and urine samples.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPyrrole-3-carboxylic acid is useful for the synthesis of cholecystokinin antagonists, benzopyran antihypertensives and azepinediones. It is utilized for the preparation of a poly(pyrrole-3-carboxylic acid) modified pencil graphite electrode and its usage in the determination of serotonin (SER) in blood serum and urine samples.
Solubility
Soluble in water.
Notes
Incompatible with strong bases, strong oxidizing agents, strong reducing agents.
Specifications
Pyrrole-3-carboxylic acid | |
C5H5NO2 | |
1 g | |
pyrrole-3-carboxylic acid, pyrrole-3-carboxylicacid, 3-carboxy-1h-pyrrole, 1h-pyrrole-3-carboxylicacid, pubchem8345, 3-carboxypyrrole, 3-pyrrolecarboxylic acid, acmc-1agqb, 3-pyrrole-carboxylic acid, pyrrole 3-carboxylic acid | |
DOYOPBSXEIZLRE-UHFFFAOYSA-N | |
1H-pyrrole-3-carboxylic acid | |
101030 | |
111.1 |
931-03-3 | |
MFCD00800594 | |
108642 | |
Soluble in water. | |
C1=CNC=C1C(=O)O | |
111.1 | |
CHEBI:68076 | |
≥98% |
Chemical Identifiers
931-03-3 | |
111.1 | |
DOYOPBSXEIZLRE-UHFFFAOYSA-N | |
101030 | |
1H-pyrrole-3-carboxylic acid |
C5H5NO2 | |
MFCD00800594 | |
pyrrole-3-carboxylic acid, pyrrole-3-carboxylicacid, 3-carboxy-1h-pyrrole, 1h-pyrrole-3-carboxylicacid, pubchem8345, 3-carboxypyrrole, 3-pyrrolecarboxylic acid, acmc-1agqb, 3-pyrrole-carboxylic acid, pyrrole 3-carboxylic acid | |
CHEBI:68076 | |
C1=CNC=C1C(=O)O |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only