Learn More
Pyrrole-2-carboxylic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1375614
Description
Pyrrole-2-carboxylic acid has demonstrated antiparasitic activity against Trypanosomes by selective proline racemase inhibition and has potent antifungal activity against Phytophthora. It also has antifungal properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPyrrole-2-carboxylic acid has demonstrated antiparasitic activity against Trypanosomes by selective proline racemase inhibition and has potent antifungal activity against Phytophthora. It also has antifungal properties.
Solubility
Soluble in methanol.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
Pyrrole-2-carboxylic acid | |
∼205°C (sublimation) | |
MFCD00005219 | |
80825 | |
Soluble in methanol. | |
C1=CNC(=C1)C(=O)O | |
111.1 | |
CHEBI:36751 | |
99% |
634-97-9 | |
C5H5NO2 | |
25 g | |
pyrrole-2-carboxylic acid, minaline, minalin, 2-pyrrolecarboxylic acid, pyrrole-2-carboxylicacid, pyrrole-2-carboxylate, 2-carboxypyrrole, 1h-pyrrolecarboxylic acid, 9ci, pyc | |
WRHZVMBBRYBTKZ-UHFFFAOYSA-N | |
1H-pyrrole-2-carboxylic acid | |
12473 | |
111.1 |
Chemical Identifiers
634-97-9 | |
111.1 | |
WRHZVMBBRYBTKZ-UHFFFAOYSA-N | |
12473 | |
1H-pyrrole-2-carboxylic acid |
C5H5NO2 | |
MFCD00005219 | |
pyrrole-2-carboxylic acid, minaline, minalin, 2-pyrrolecarboxylic acid, pyrrole-2-carboxylicacid, pyrrole-2-carboxylate, 2-carboxypyrrole, 1h-pyrrolecarboxylic acid, 9ci, pyc | |
CHEBI:36751 | |
C1=CNC(=C1)C(=O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 211-221-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only