Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyrrole-2-carboxaldoxime, 97%

$36.61 - $415.86

Chemical Identifiers

Chemical Name or Material Pyrrole-2-carboxaldoxime
CAS 32597-34-5
Molecular Formula C5H6N2O
Formula Weight 110.12
MDL Number MFCD00965095
Beilstein 108674
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1895903

  SDS
alfa aesar™
L1895903
1g Each for $36.61

AAL1895906

  SDS
alfa aesar™
L1895906
5g Each for $108.18

AAL1895914

  SDS
alfa aesar™
L1895914
25g Each for $415.86
Description & Specifications

Specifications

Formula Weight 110.12
CAS 32597-34-5
Chemical Name or Material Pyrrole-2-carboxaldoxime
MDL Number MFCD00965095
Quantity 1g
Assay 97%
Melting Point 163° to 166°C
Beilstein 108674
Molecular Formula C5H6N2O