missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrrole-2-carboxaldehyde 98.0+%, TCI America™
Supplier: TCI America P12465G
Specifications
| Pyrrole-2-carboxaldehyde | |
| 45°C | |
| C5H5NO | |
| 5 g | |
| ZSKGQVFRTSEPJT-UHFFFAOYSA-N | |
| 1H-pyrrole-2-carbaldehyde | |
| 13854 | |
| 95.10 | |
| Crystalline Powder |
| 1003-29-8 | |
| 219°C | |
| MFCD00005217 | |
| pyrrole-2-carboxaldehyde, 2-formylpyrrole, 1h-pyrrole-2-carboxaldehyde, pyrrole-2-aldehyde, 2-pyrrolecarboxaldehyde, pyrrole-2-carbaldehyde, 2-pyrrolecarbaldehyde, 2-pyrrolylcarboxaldehyde, 2-pyrrolaldehyde, alpha-pyrrolaldehyde | |
| C1=CNC(=C1)C=O | |
| 95.101 | |
| CHEBI:59978 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 1003-29-8 | |
| 95.101 | |
| ZSKGQVFRTSEPJT-UHFFFAOYSA-N | |
| 13854 | |
| 1H-pyrrole-2-carbaldehyde |
| C5H5NO | |
| MFCD00005217 | |
| pyrrole-2-carboxaldehyde, 2-formylpyrrole, 1h-pyrrole-2-carboxaldehyde, pyrrole-2-aldehyde, 2-pyrrolecarboxaldehyde, pyrrole-2-carbaldehyde, 2-pyrrolecarbaldehyde, 2-pyrrolylcarboxaldehyde, 2-pyrrolaldehyde, alpha-pyrrolaldehyde | |
| CHEBI:59978 | |
| C1=CNC(=C1)C=O |
Safety and Handling
TSCA : Yes