Learn More
Pyrrole-2-carboxaldehyde, 99%, Thermo Scientific Chemicals
Pyrrole-2-carboxaldehyde, 99%, C5H5NO, CAS Number-1003-29-8, 2-pyrrolecarboxaldehyde, 2-pyrrolecarbaldehyde, 2-pyrrolaldehyde, 1h-pyrrole-2-carboxaldehyde, alpha-pyrrolaldehyde, 2-formylpyrrole, 2-pyrrolylcarboxaldehyde, pyrrole-2-aldehyde, pyrrole-2-carboxaldehyde, pyrrole-2-carbaldehyde, 100g
$126.92 - $799.47
Chemical Identifiers
CAS | 1003-29-8 |
---|---|
Molecular Formula | C5H5NO |
Molecular Weight (g/mol) | 95.101 |
MDL Number | MFCD00005217 |
InChI Key | ZSKGQVFRTSEPJT-UHFFFAOYSA-N |
Synonym | pyrrole-2-carboxaldehyde, 2-formylpyrrole, 1h-pyrrole-2-carboxaldehyde, pyrrole-2-aldehyde, 2-pyrrolecarboxaldehyde, pyrrole-2-carbaldehyde, 2-pyrrolecarbaldehyde, 2-pyrrolylcarboxaldehyde, 2-pyrrolaldehyde, alpha-pyrrolaldehyde |
PubChem CID | 13854 |
ChEBI | CHEBI:59978 |
IUPAC Name | 1H-pyrrole-2-carbaldehyde |
SMILES | C1=CNC(=C1)C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1283809
|
Thermo Scientific Chemicals
A1283809 |
10 g |
Each for $126.92
|
|
|||||
AAA1283814
|
Thermo Scientific Chemicals
A1283814 |
25 g |
Each for $277.78
|
|
|||||
AAA1283822
|
Thermo Scientific Chemicals
A1283822 |
100 g |
Each for $799.47
|
|
|||||
Description
Pyrrole-2-carboxaldehyde is used as a pharmaceutical intermediate. It undergoes condensation reaction to produce pyrrole-2-carboxaldehyde salicylhydrazone (PCSH). It is used as a key starting material for the preparation of pyrrole -2-carboxylic acid by oxidation reaction. It is also used to prepare 4-Difluoro-4-bora-3a,4a-diaza-s-indacene (BODIPY dyes) through condensation with a pyrrole, which is an important probes for biotechnology and other applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPyrrole-2-carboxaldehyde is used as a pharmaceutical intermediate. It undergoes condensation reaction to produce pyrrole-2-carboxaldehyde salicylhydrazone (PCSH). It is used as a key starting material for the preparation of pyrrole -2-carboxylic acid by oxidation reaction. It is also used to prepare 4-Difluoro-4-bora-3a,4a-diaza-s-indacene (BODIPY dyes) through condensation with a pyrrole, which is an important probes for biotechnology and other applications.
Solubility
Soluble in chloroform, dimethyl sulfoxide and methanol. Insoluble in water.
Notes
Store in cool place. Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
Chemical Identifiers
1003-29-8 | |
95.101 | |
ZSKGQVFRTSEPJT-UHFFFAOYSA-N | |
13854 | |
1H-pyrrole-2-carbaldehyde |
C5H5NO | |
MFCD00005217 | |
pyrrole-2-carboxaldehyde, 2-formylpyrrole, 1h-pyrrole-2-carboxaldehyde, pyrrole-2-aldehyde, 2-pyrrolecarboxaldehyde, pyrrole-2-carbaldehyde, 2-pyrrolecarbaldehyde, 2-pyrrolylcarboxaldehyde, 2-pyrrolaldehyde, alpha-pyrrolaldehyde | |
CHEBI:59978 | |
C1=CNC(=C1)C=O |
Specifications
1003-29-8 | |
217°C to 219°C | |
C5H5NO | |
10 g | |
Air sensitive | |
Soluble in chloroform,dimethyl sulfoxide and methanol. Insoluble in water. | |
C1=CNC(=C1)C=O | |
95.101 | |
CHEBI:59978 | |
99% |
40°C to 46°C | |
106°C (222°F) | |
MFCD00005217 | |
105745 | |
pyrrole-2-carboxaldehyde, 2-formylpyrrole, 1h-pyrrole-2-carboxaldehyde, pyrrole-2-aldehyde, 2-pyrrolecarboxaldehyde, pyrrole-2-carbaldehyde, 2-pyrrolecarbaldehyde, 2-pyrrolylcarboxaldehyde, 2-pyrrolaldehyde, alpha-pyrrolaldehyde | |
ZSKGQVFRTSEPJT-UHFFFAOYSA-N | |
1H-pyrrole-2-carbaldehyde | |
13854 | |
95.1 | |
Pyrrole-2-carboxaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 213-705-5
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only