Please login to your online account to display your discounted pricing

Pyroxamide, Tocris™

Histone deacetylase inhibitor

$218.74 - $915.10

Chemical Identifiers

Chemical Name or Material Pyroxamide
Assay Percent Range >98%
Molecular Formula C13H19N3O3
Formula Weight 265.31
Synonym N-Hydroxy-N'-3-pyridinyloctanediamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

440310

 
r & d systems
RDS440310
10mg Each for $218.74

440350

 
r & d systems
RDS440350
50mg Each for $915.10
Description & Specifications

Specifications

Formula Weight 265.31
Quantity 10mg
Assay Percent Range >98%
Molecular Formula C13H19N3O3
CAS 382180-17-8
Synonym N-Hydroxy-N'-3-pyridinyloctanediamide
Chemical Name or Material Pyroxamide
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C