Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyrophosphoryl chloride

Chemical Identifiers

Chemical Name or Material Pyrophosphoryl chloride
CAS 13498-14-1
Formula Weight 251.76
MDL Number MFCD00061542
UN Number UN3264
Synonym Phosphoryl chloride, pyro
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA2236202

  SDS
alfa aesar™
2236202
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA2236206

  SDS
alfa aesar™
2236206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA2236214

  SDS
alfa aesar™
2236214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 251.76
Chemical Name or Material Pyrophosphoryl chloride
Odor Acrid
Physical Form Liquid
MDL Number MFCD00061542
CAS 13498-14-1
UN Number UN3264
Density 1.82g/mL
Synonym Phosphoryl chloride, pyro
Color Colorless to pale yellow
Solubility Information Reacts violently with water
Melting Point −17°C
Boiling Point 101°C (10mmHg)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
Sensitivity Air & Moisture Sensitive
Quantity 0.5g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.