missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyrophosphoryl chloride

Manufacturer:  Alfa Aesar™ 2236214

 View more versions of this product

Catalog No. AA2236214



Specifications

Specifications

Pyrophosphoryl chloride
Acrid
101°C (10mmHg)
-17°C
13498-14-1
MFCD00061542
diphosphoryl chloride, pyrophosphoryl chloride, diphosphoryl tetrachloride, diphosphoric tetrachloride, pyrophosphoric tetrachloride, dichlorophosphoryl oxyphosphonoyl dichloride, pyrophosphoryl tetrachloride, pyrophosphorylchloride, diphosphoryl chloride, phosphorodichloridic anhydride
CNTIXUGILVWVHR-UHFFFAOYSA-N
251.745
251.76
1.82g/mL
Air & Moisture Sensitive
Colorless to Yellow
25g
Cl4O3P2
UN3264
Reacts violently with water
O=P(OP(=O)(Cl)Cl)(Cl)Cl
4432411
Liquid

Chemical Identifiers

13498-14-1
251.745
CNTIXUGILVWVHR-UHFFFAOYSA-N
4432411
Cl4O3P2
MFCD00061542
diphosphoryl chloride, pyrophosphoryl chloride, diphosphoryl tetrachloride, diphosphoric tetrachloride, pyrophosphoric tetrachloride, dichlorophosphoryl oxyphosphonoyl dichloride, pyrophosphoryl tetrachloride, pyrophosphorylchloride, diphosphoryl chloride, phosphorodichloridic anhydride
O=P(OP(=O)(Cl)Cl)(Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

SDS Documents