Please login to your online account to display your discounted pricing

Pyronin Y, pure, high purity biological stain, ACROS Organics™

$156.83 - $672.55

Chemical Identifiers

Chemical Name or Material Pyronin Y
Name Note Pure
CAS 92-32-0
Purity Grade Pure
Molecular Formula C17H19ClN2O
Formula Weight 302.79
MDL Number MFCD00011725
Synonym C.I. 45005
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC200100250

  SDS
thermo fisher scientific
AC200100250
Glass bottle 25g Each for $672.55

AC200100010

  SDS
fisher scientific acros
200100010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC200100050

  SDS
thermo fisher scientific
200100050
Glass bottle 5g Each for $156.83
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 302.79
Quantity 25g
Color Dark green
Purity Grade Pure
Merck Index 15, 8119
Beilstein 18, V,10, 181
Chemical Name or Material Pyronin Y
Synonym C.I. 45005
Name Note Pure
Molecular Formula C17H19ClN2O
CAS 92-32-0
MDL Number MFCD00011725
Dye Content 50% min.
Packaging Glass bottle
Absorption Ratio 1.23 to 1.7
Lambda Max 546 to 551nm (in 50% ethanol)
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Suspected of causing genetic defects.
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Suspected of causing genetic defects.