missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyronin Y, 0.2% w/v aq. soln.

$48.89 - $72.93

Chemical Identifiers

CAS 92-32-0
Molecular Formula C17H19ClN2O
Molecular Weight (g/mol) 302.802
MDL Number MFCD00011725
InChI Key INCIMLINXXICKS-UHFFFAOYSA-M
Synonym C.I. 45005; Pyronin G
PubChem CID 7085
ChEBI CHEBI:87347
IUPAC Name [6-(dimethylamino)xanthen-3-ylidene]-dimethylazanium;chloride
SMILES CN(C)C1=CC2=C(C=C1)C=C3C=CC(=[N+](C)C)C=C3O2.[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60505AB View Documents Alfa Aesar™
J60505AB
10mL Each for $48.89
 
AAJ60505AC View Documents Alfa Aesar™
J60505AC
25mL Each for $72.93
 
Specifications

Chemical Identifiers

92-32-0
302.802
INCIMLINXXICKS-UHFFFAOYSA-M
7085
[6-(dimethylamino)xanthen-3-ylidene]-dimethylazanium;chloride
C17H19ClN2O
MFCD00011725
C.I. 45005; Pyronin G
CHEBI:87347
CN(C)C1=CC2=C(C=C1)C=C3C=CC(=[N+](C)C)C=C3O2.[Cl-]

Specifications

92-32-0
C17H19ClN2O
3795789
C.I. 45005; Pyronin G
CN(C)C1=CC2=C(C=C1)C=C3C=CC(=[N+](C)C)C=C3O2.[Cl-]
302.802
CHEBI:87347
Liquid
Pink
Pyronin Y
0.2% w/v aq. soln.
MFCD00011725
14,8007
INCIMLINXXICKS-UHFFFAOYSA-M
[6-(dimethylamino)xanthen-3-ylidene]-dimethylazanium;chloride
7085
302.8
Odorless
10mL
Safety and Handling

Safety and Handling

TSCA : TSCA