Learn More
Pyrogallol, ACS, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04415218
Description
Pyrogallol is used in hair dying and suturing materials dying. It is used as a developing agent in black and white developers, as a metal- complexing agent and is involved in oxygen absorption in gas analysis. It exhibits antiseptic properties. Its alkaline solution is used to absorb oxygen from air, thereby it plays a vital role to calculate the amount of oxygen in air.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Pyrogallol | |
131°C to 135°C | |
1.45 g/mL | |
Odorless | |
MFCD00002192 | |
UN2811 | |
Air and light sensitive | |
pyrogallol, 1,2,3-trihydroxybenzene, pyrogallic acid, 1,2,3-benzenetriol, fouramine brown ap, fourrine pg, fourrine 85, pyro, c.i. oxidation base 32, piral | |
WQGWDDDVZFFDIG-UHFFFAOYSA-N | |
benzene-1,2,3-triol | |
1057 | |
126.11 | |
ACS |
87-66-1 | |
White | |
309°C | |
C6H6O3 | |
50 g | |
907431 | |
14,8000 | |
Soluble in ethanol,water and ether. Slightly soluble in chloroform. | |
OC1=CC=CC(O)=C1O | |
126.11 | |
CHEBI:16164 | |
99% | |
Powder |
Chemical Identifiers
87-66-1 | |
126.11 | |
WQGWDDDVZFFDIG-UHFFFAOYSA-N | |
1057 | |
benzene-1,2,3-triol |
C6H6O3 | |
MFCD00002192 | |
pyrogallol, 1,2,3-trihydroxybenzene, pyrogallic acid, 1,2,3-benzenetriol, fouramine brown ap, fourrine pg, fourrine 85, pyro, c.i. oxidation base 32, piral | |
CHEBI:16164 | |
OC1=CC=CC(O)=C1O |
Safety and Handling
GHS H Statement
H341-H302-H312-H332
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
H301-H312+H332-H315-H319-H341
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 201-762-9
RTECSNumber : UX2800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only