Learn More
Pyrogallol, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 164560025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Pyrogallol | |
309.0°C | |
98.5% min. (HPLC) | |
C6H6O3 | |
MFCD00002192 | |
06,1071 | |
pyrogallol, 1,2,3-trihydroxybenzene, pyrogallic acid, 1,2,3-benzenetriol, fouramine brown ap, fourrine pg, fourrine 85, pyro, c.i. oxidation base 32, piral | |
WQGWDDDVZFFDIG-UHFFFAOYSA-N | |
benzene-1,2,3-triol | |
1057 | |
126.11 |
87-66-1 | |
Authentic | |
Plastic bottle | |
C6H3(OH)3 | |
2.5kg | |
15,8112 | |
Solubility in water: 400g/L (25°C). Other solubilities: 1 g/1.3 mL alcohol, 1 g/1.6 mL ether, slightly soluble in benzene, chloroform and, carbon disulfide | |
OC1=CC=CC(O)=C1O | |
126.11 | |
CHEBI:16164 | |
99% |
Safety and Handling
GHS H Statement
Harmful to aquatic life with long lasting effects.
Toxic if swallowed.
Suspected of causing genetic defects.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Causes serio
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If eye irr
GHS Signal Word: Danger
EINECSNumber : 201-762-9
RUO â Research Use Only