missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyrimidine, 99%, ACROS Organics™

Manufacturer:  Acros Organics 157700050

 View more versions of this product

Catalog No. AC157700050



Specifications

Specifications

Pyrimidine
Authentic
1.5010 to 1.5040 (20°C, 589nm)
5g
99%
MFCD00006059
15, 8100
CZPWVGJYEJSRLH-UHFFFAOYSA-N
pyrimidine
9260
80.09
≥98.5%
1.01g/mL
Glass bottle
Colorless to Orange
289-95-2
C4H4N2
23, 89
1,3-diazine, metadiazine, m-diazine, miazine, 1,3-diazabenzene, base, 1,3-diazin, pyrimidin, unii-k8cxk5q32l, k8cxk5q32l
C1=CN=CN=C1
80.09
CHEBI:16898
Liquid

Chemical Identifiers

289-95-2
80.09
CZPWVGJYEJSRLH-UHFFFAOYSA-N
9260
pyrimidine
C4H4N2
MFCD00006059
1,3-diazine, metadiazine, m-diazine, miazine, 1,3-diazabenzene, base, 1,3-diazin, pyrimidin, unii-k8cxk5q32l, k8cxk5q32l
CHEBI:16898
C1=CN=CN=C1
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents