missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyrimidine-2-thiocarboxamide, 97%

Manufacturer:  Alfa Aesar™ H54783MD

 View more versions of this product

Catalog No. AAH54783MD



Description

Description

Applications
Pyrimidine-2-thiocarboxamide is used as a pharmaceutical intermediate and as a synthetic building block.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

Pyrimidine-2-thiocarboxamide
4537-73-9
MFCD12093703
2-pyrimidinecarbothioamide, 2-pyrimidinecarbothioamide 9ci, pyrimidine-2-thiocarboxamide
C1=CN=C(N=C1)C(=S)N
139.176
139.17
250mg
C5H5N3S
508162
QAUHVPUYFSGVME-UHFFFAOYSA-N
pyrimidine-2-carbothioamide
23273397
97%

Chemical Identifiers

4537-73-9
139.176
QAUHVPUYFSGVME-UHFFFAOYSA-N
23273397
C1=CN=C(N=C1)C(=S)N
C5H5N3S
MFCD12093703
2-pyrimidinecarbothioamide, 2-pyrimidinecarbothioamide 9ci, pyrimidine-2-thiocarboxamide
pyrimidine-2-carbothioamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents