missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyrimidine-2-carboxylic acid, 97%

Manufacturer:  Alfa Aesar™ H5474503

 View more versions of this product

Catalog No. AAH5474503



Description

Description

Applications
2-carboxy-Pyrimidine is a synthetic intermediate.

Solubility
Soluble in ethanol, Dimethyl sulfoxide (DMSO), Dimethylformamide (DMF), and solution of Phosphate Buffered Saline (PBS).

Notes
Store at -20°C.
Specifications

Specifications

Pyrimidine-2-carboxylic acid
31519-62-7
MFCD00856161
Soluble in ethanol,Dimethyl sulfoxide (DMSO),Dimethylformamide (DMF),and solution of Phosphate Buffered Saline (PBS).
C1=CN=C(N=C1)C(=O)O
124.099
124.1
1g
C5H4N2O2
2-carboxy pyrimidine
ZFCHNZDUMIOWFV-UHFFFAOYSA-N
pyrimidine-2-carboxylic acid
12626245
97%

Chemical Identifiers

31519-62-7
124.099
ZFCHNZDUMIOWFV-UHFFFAOYSA-N
12626245
C1=CN=C(N=C1)C(=O)O
C5H4N2O2
MFCD00856161
2-carboxy pyrimidine
pyrimidine-2-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents