missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyrimidine-2-carboxamidoxime, 97%

Manufacturer:  Alfa Aesar™ H5435106

 View more versions of this product

Catalog No. AAH5435106



Specifications

Specifications

Pyrimidine-2-carboxamidoxime
5g
C5H6N4O
2-pyrimidinecarboxamide oxime
C1=CNC(=C(N)N=O)N=C1
138.13
138.13
230°C to 235°C
90993-49-0
MFCD09751609
RYYZBKPHKBOVSQ-UHFFFAOYSA-N
nitroso(1H-pyrimidin-2-ylidene)methanamine
51550622
97%

Chemical Identifiers

90993-49-0
138.13
RYYZBKPHKBOVSQ-UHFFFAOYSA-N
51550622
C1=CNC(=C(N)N=O)N=C1
C5H6N4O
MFCD09751609
2-pyrimidinecarboxamide oxime
nitroso(1H-pyrimidin-2-ylidene)methanamine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents