missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyrilamine Maleate, TCI America™

Manufacturer:  TCI America P23695G

 View more versions of this product

Catalog No. P23695G



Specifications

Specifications

Pyrilamine Maleate
59-33-6
C21H27N3O5
Mepyramine Maleate, N-(4-Methoxyphenyl)methyl-N′C,N′C-dimethyl-N-(2-pyridinyl)-1,2-ethanediamine Maleate
JXYWFNAQESKDNC-BTJKTKAUSA-N
CN(C)CCN(CC1=CC=C(C=C1)OC)C2=CC=CC=N2.C(=CC(=O)O)C(=O)O
401.463
401.46
5g
≥98.0% (HPLC,T)
MFCD00069333
White
102°C
(Z)-but-2-enedioic acid;N'-[(4-methoxyphenyl)methyl]-N,N-dimethyl-N'-pyridin-2-ylethane-1,2-diamine
5284451
Crystal-Powder at 20°C

Chemical Identifiers

59-33-6
401.463
JXYWFNAQESKDNC-BTJKTKAUSA-N
5284451
CN(C)CCN(CC1=CC=C(C=C1)OC)C2=CC=CC=N2.C(=CC(=O)O)C(=O)O
C21H27N3O5
MFCD00069333
Mepyramine Maleate, N-(4-Methoxyphenyl)methyl-N′C,N′C-dimethyl-N-(2-pyridinyl)-1,2-ethanediamine Maleate
(Z)-but-2-enedioic acid;N'-[(4-methoxyphenyl)methyl]-N,N-dimethyl-N'-pyridin-2-ylethane-1,2-diamine
Safety and Handling

Safety and Handling

RTECSNumber : UT1225000

TSCA : Yes

Documents