missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyrilamine maleate

Selective inverse agonist for the H-1 histamine receptor

$161.92 - $161.92

Chemical Identifiers

CAS 59-33-6
Molecular Formula C21H27N3O5
Molecular Weight (g/mol) 401.463
MDL Number MFCD00069333
InChI Key JXYWFNAQESKDNC-BTJKTKAUSA-N
Synonym pyrilamine maleate, mepyramine maleate, anisopyradamine, histosol, minihist, diaminide maleate, paraminyl maleate, antihist, histatex, histine
PubChem CID 5284451
IUPAC Name (Z)-but-2-enedioic acid;N'-[(4-methoxyphenyl)methyl]-N,N-dimethyl-N'-pyridin-2-ylethane-1,2-diamine
SMILES CN(C)CCN(CC1=CC=C(C=C1)OC)C2=CC=CC=N2.C(=CC(=O)O)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6324714 View Documents Alfa Aesar™
J6324714
25g Each for $161.92
 
Description

Description

Applications
Pyrilamine maleate Selective inverse agonist for the H-1 histamine receptor. Pyrilamine Maleate is commonly utilized as a radioligand ([3H]Mepyramine) binding assay for the H1 receptor. And also used to exhibit blocking ability of KCNQ/M channels which is potentially related to an adverse effect seen in excess intake of antihistamines.

Solubility
Soluble in water at 100mg/ml

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

59-33-6
401.463
JXYWFNAQESKDNC-BTJKTKAUSA-N
5284451
CN(C)CCN(CC1=CC=C(C=C1)OC)C2=CC=CC=N2.C(=CC(=O)O)C(=O)O
C21H27N3O5
MFCD00069333
pyrilamine maleate, mepyramine maleate, anisopyradamine, histosol, minihist, diaminide maleate, paraminyl maleate, antihist, histatex, histine
(Z)-but-2-enedioic acid;N'-[(4-methoxyphenyl)methyl]-N,N-dimethyl-N'-pyridin-2-ylethane-1,2-diamine

Specifications

White
59-33-6
MFCD00069333
pyrilamine maleate, mepyramine maleate, anisopyradamine, histosol, minihist, diaminide maleate, paraminyl maleate, antihist, histatex, histine
JXYWFNAQESKDNC-BTJKTKAUSA-N
(Z)-but-2-enedioic acid;N'-[(4-methoxyphenyl)methyl]-N,N-dimethyl-N'-pyridin-2-ylethane-1,2-diamine
5284451
Powder
25g
C21H27N3O5
14,7984
Soluble in water at 100mg/ml
CN(C)CCN(CC1=CC=C(C=C1)OC)C2=CC=CC=N2.C(=CC(=O)O)C(=O)O
401.463
401.46
Pyrilamine maleate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UT1225000

TSCA : TSCA