missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyridoxine Hydrochloride, U.S.P. - F.C.C., J.T.Baker™

$142.70 - $1,534.67

Chemical Identifiers

CAS 58-56-0
Molecular Formula C8H11NO3·HCl
Formula Weight 205.64
Chemical Name or Material Pyridoxine Hydrochloride
Grade FCC/USP
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
02003934 Avantor Performance
334305
Wide Mouth Amber Glass Bottle 100g Each for $1,534.67
02003935 Avantor Performance
334301
Wide Mouth Amber Glass Bottle 5g Each for $142.70
Specifications Description & Specifications

Chemical Identifiers

58-56-0
205.64
FCC/USP
C8H11NO3·HCl
Pyridoxine Hydrochloride

Specifications

100g
Wide Mouth Amber Glass Bottle
205.64
FCC/USP
58-56-0
C8H11NO3·HCl
Pyridoxine Hydrochloride