Please login to your online account to display your discounted pricing

Pyridoxine Hydrochloride, Fisher BioReagents


Quantity 50g
Packaging Amber Glass

Manufacturer: fisher scientific company  BP267750

 View more versions of this product

Catalog No. BP267750

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Melting Point 214°C
Physical Form Powder/Solid
Formula Weight 205.64
Quantity 50g
Molecular Formula C8H11NO3 · HCl
pH 2.4 to 3.0
Color White
Merck Index 15, 8095
Chloride 16.9 to 17.6%
Identification Pass Test
Ignition Residue 0.1% max.
Synonym Pyridoxol Hydrochloride
Recommended Storage RT, protect from light.
CAS 58-56-0
ChemAlert Storage Symbol Gray
Packaging Amber Glass
Loss on Drying 0.5% max.
Purity 98% min.
Heavy Metals (as Pb) 0.003% max.
Infrared Spectrum Conforms to reference
Assay Percent Range ≥98 %
Chemical Name or Material Pyridoxine Hydrochloride

Specifications

Chemical Name or Material Pyridoxine Hydrochloride
CAS 58-56-0
Physical Form Powder/Solid
Assay Percent Range ≥98 %
Molecular Formula C8H11NO3 · HCl
Formula Weight 205.64
Synonym Pyridoxol Hydrochloride
Accessories
 

RT, protect from light.

SDS & Safety Documents