missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyridoxine Hydrochloride, Fisher BioReagents

$123.27 - $386.68

Chemical Identifiers

CAS 58-56-0
Molecular Formula C8H12ClNO3
Molecular Weight (g/mol) 205.638
InChI Key ZUFQODAHGAHPFQ-UHFFFAOYSA-N
Synonym pyridoxine hydrochloride, pyridoxine hcl, pyridoxol hydrochloride, vitamin b6, alestrol, becilan, benadon, hexavibex, hexermin, hexobion
PubChem CID 6019
ChEBI CHEBI:30961
IUPAC Name 4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride
SMILES CC1=NC=C(C(=C1O)CO)CO.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP267750 SDS View Documents Fisher BioReagents
BP267750
50g Amber Glass Each for $214.71
Case of 6 Each for $1,288.26
BP267710 SDS View Documents Fisher BioReagents
BP267710
10g Amber Glass Each for $123.27
Case of 7 Each for $862.89
BP2677100 SDS View Documents Fisher BioReagents
BP2677100
100g Amber Glass Each for $386.68
Case of 3 Each for $1,160.04
Description

Description

Pyridoxine HCl is used as a vitamin (enzyme cofactor).
Specifications

Chemical Identifiers

58-56-0
205.638
pyridoxine hydrochloride, pyridoxine hcl, pyridoxol hydrochloride, vitamin b6, alestrol, becilan, benadon, hexavibex, hexermin, hexobion
CHEBI:30961
CC1=NC=C(C(=C1O)CO)CO.Cl
C8H12ClNO3
ZUFQODAHGAHPFQ-UHFFFAOYSA-N
6019
4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride

Specifications

Pass Test
Conforms to reference
Amber Glass
214°C
2.4 to 3.0
≥98 %
15, 8095
ZUFQODAHGAHPFQ-UHFFFAOYSA-N
4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride
6019
205.64
98% min.
0.1% max.
0.5% max.
White
50g
58-56-0
C8H12ClNO3
pyridoxine hydrochloride, pyridoxine hcl, pyridoxol hydrochloride, vitamin b6, alestrol, becilan, benadon, hexavibex, hexermin, hexobion
CC1=NC=C(C(=C1O)CO)CO.Cl
205.638
CHEBI:30961
Powder/Solid
Pyridoxine Hydrochloride
Safety and Handling

Safety and Handling

Recommended Storage : RT, protect from light.