Please login to your online account to display your discounted pricing

Pyridoxine Hydrochloride, Fisher BioReagents


Quantity 100g
Packaging Amber Glass

Manufacturer: fisher scientific company  BP2677100

 View more versions of this product

Catalog No. BP2677100

  • / Each
  • / Case of 3 Each
 

Description & Specifications

Chemical Identifiers

Melting Point 214°C
Formula Weight 205.64
Physical Form Powder/Solid
Quantity 100g
Merck Index 15, 8095
Molecular Formula C8H11NO3 · HCl
Color White
pH 2.4 to 3.0
Heavy Metals (as Pb) 0.003% max.
Chloride 16.9 to 17.6%
Ignition Residue 0.1% max.
Recommended Storage RT, protect from light.
Loss on Drying 0.5% max.
Assay Percent Range ≥98 %
Infrared Spectrum Conforms to reference
Synonym Pyridoxol Hydrochloride
Purity 98% min.
Identification Pass Test
Packaging Amber Glass
Chemical Name or Material Pyridoxine Hydrochloride
ChemAlert Storage Symbol Gray
CAS 58-56-0

Specifications

Chemical Name or Material Pyridoxine Hydrochloride
CAS 58-56-0
Physical Form Powder/Solid
Assay Percent Range ≥98 %
Molecular Formula C8H11NO3 · HCl
Formula Weight 205.64
Synonym Pyridoxol Hydrochloride
Accessories
 

RT, protect from light.

SDS & Safety Documents