Please login to your online account to display your discounted pricing

Pyridoxine Hydrochloride, Fisher BioReagents


Quantity 10g
Packaging Amber Glass

Manufacturer: fisher scientific company  BP267710

 View more versions of this product

Catalog No. BP267710

  • / Each
  • / Case of 7 Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder/Solid
Formula Weight 205.64
Melting Point 214°C
Quantity 10g
Merck Index 15, 8095
Molecular Formula C8H11NO3 · HCl
Vapor Density 7.1g/mol
Color White
pH 2.4 to 3.0
Chemical Name or Material Pyridoxine Hydrochloride
Loss on Drying 0.5% max.
Packaging Amber Glass
Purity 98% min.
Chloride 16.9 to 17.6%
Synonym Pyridoxol Hydrochloride
Infrared Spectrum Conforms to reference
Ignition Residue 0.1% max.
Assay Percent Range ≥98 %
CAS 58-56-0
ChemAlert Storage Symbol Gray
Heavy Metals (as Pb) 0.003% max.
Identification Pass Test
Recommended Storage RT, protect from light.

Specifications

Chemical Name or Material Pyridoxine Hydrochloride
CAS 58-56-0
Physical Form Powder/Solid
Assay Percent Range ≥98 %
Molecular Formula C8H11NO3 · HCl
Formula Weight 205.64
Synonym Pyridoxol Hydrochloride
Accessories
 

RT, protect from light.

SDS & Safety Documents