missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyridoxine Dicaprylate, TCI America™

Manufacturer:  TCI America P220825G

 View more versions of this product

Catalog No. P220825G



Specifications

Specifications

Pyridoxine Dicaprylate
72°C
106483-04-9
PAUSGZCRNOTKPK-UHFFFAOYSA-N
[5-hydroxy-6-methyl-4-(octanoyloxymethyl)pyridin-3-yl]methyl octanoate
83816
Crystal-Powder at 20°C
White
25g
C24H39NO5
CCCCCCCC(=O)OCC1=CN=C(C(=C1COC(=O)CCCCCCC)O)C
421.578
421.58
≥98.0% (N)

Chemical Identifiers

106483-04-9
421.578
83816
CCCCCCCC(=O)OCC1=CN=C(C(=C1COC(=O)CCCCCCC)O)C
C24H39NO5
PAUSGZCRNOTKPK-UHFFFAOYSA-N
[5-hydroxy-6-methyl-4-(octanoyloxymethyl)pyridin-3-yl]methyl octanoate
Safety and Handling

Safety and Handling

EINECSNumber : (9)-2209

TSCA : No

Documents