missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pyridoxamine dihydrochloride, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC436250050
Specifications
Pyridoxamine dihydrochloride | |
Authentic | |
10% Max. (K.F.) | |
216.0°C to 218.0°C | |
524-36-7 | |
C8H12N2O2·2ClH | |
2-methyl-3-hydroxy-4-aminomethyl-5-hydroxymethylpyridene dihydrochloride, 4-aminomethyl-5-hydroxy-6-methyl-3-pyridinemethanol dihydrochloride, 4-aminomethyl-5-hydroxymethyl-2-methylpyridin-3-ol dihydrochloride, pyridoxamine 2hcl, pyridoxamine dichlorohydrate, pyridoxamine dihydrochloride, pyridoxemine, dihydrochloride, pyridoxylamine dihydrochloride, unii-yq8ncr7v0o, yq8ncr7v0o | |
CC1=NC=C(C(=C1O)CN)CO.Cl.Cl | |
241.12 | |
241.12 | |
98% |
36.15 to 38.01% C, 10.54 to 11.08%N (based on monohydrate) | |
Glass bottle | |
Brown to White | |
5g | |
97.5 to max. 102.5% min. (on dry basis) (Argentometry) | |
15,8093 | |
HNWCOANXZNKMLR-UHFFFAOYSA-N | |
4-(aminomethyl)-5-(hydroxymethyl)-2-methylpyridin-3-ol;dihydrochloride | |
10664 | |
Powder or Solid |
Chemical Identifiers
524-36-7 | |
241.12 | |
2-methyl-3-hydroxy-4-aminomethyl-5-hydroxymethylpyridene dihydrochloride, 4-aminomethyl-5-hydroxy-6-methyl-3-pyridinemethanol dihydrochloride, 4-aminomethyl-5-hydroxymethyl-2-methylpyridin-3-ol dihydrochloride, pyridoxamine 2hcl, pyridoxamine dichlorohydrate, pyridoxamine dihydrochloride, pyridoxemine, dihydrochloride, pyridoxylamine dihydrochloride, unii-yq8ncr7v0o, yq8ncr7v0o | |
4-(aminomethyl)-5-(hydroxymethyl)-2-methylpyridin-3-ol;dihydrochloride |
C8H12N2O2·2ClH | |
HNWCOANXZNKMLR-UHFFFAOYSA-N | |
10664 | |
CC1=NC=C(C(=C1O)CN)CO.Cl.Cl |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
EINECSNumber : 208-357-6