missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyridoxal Hydrochloride, Fisher BioReagents

Manufacturer:  Fisher Scientific Company BP26755

 View more versions of this product

Catalog No. BP26755



 

Specifications Description & Specifications

Specifications

203.63
171°C
C8H9NO3 · HCl
2.1
65-22-5
Conforms to reference
Pass Test
Powder/Solid
5g
Off-white
15, 8091
Pyridoxal Hydrochloride
Amber Glass
253-5, 290-1, 316-8nm

Chemical Identifiers

65-22-5
C8H9NO3 · HCl
Pyridoxal Hydrochloride
Powder/Solid
203.63

Safety and Handling

SDS & Safety Documents

Recommended Storage : 0°C, desiccate, protect from light.