Please login to your online account to display your discounted pricing

Pyridoxal Hydrochloride, Fisher BioReagents

$119.85 - $183.88

Chemical Identifiers

Chemical Name or Material Pyridoxal Hydrochloride
CAS 65-22-5
Physical Form Powder/Solid
Molecular Formula C8H9NO3 · HCl
Formula Weight 203.63
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BP26751

  SDS
fisher scientific company
BP26751
Amber Glass 1g Each for $119.85
Case of 9 Each for $1,078.65

BP26755

  SDS
fisher scientific company
BP26755
Amber Glass 5g Each for $183.88
Case of 9 Each for $1,654.92
Description & Specifications

Specifications

Physical Form Powder/Solid
Formula Weight 203.63
Melting Point 171°C
Quantity 1g
Merck Index 15, 8091
Molecular Formula C8H9NO3 · HCl
Color Off-white
pH 2.1
Packaging Amber Glass
FTIR Conforms to reference
Chemical Name or Material Pyridoxal Hydrochloride
CAS 65-22-5
ChemAlert Storage Symbol Gray
Identification Pass Test
Recommended Storage 0°C, desiccate, protect from light.
Lambda Max 253-5, 290-1, 316-8nm

Pyridoxal Hydrochloride is useful for labeling amino acids and for their detection in the picomole range.

Safety and Handling
 

0°C, desiccate, protect from light.