missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pyridoxal hydrochloride, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 352710010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Pyridoxal hydrochloride | |
171.0°C to 175.0°C | |
Authentic | |
Glass bottle | |
MFCD00012809 | |
15,8091 | |
Solubility in water: 500g/L (20°C). Other solubilities: soluble in 95% ethanol: 1.7g/100 mL | |
CC1=NC=C(C(=C1O)C=O)CO.Cl | |
203.62 | |
203.62 | |
Powder |
65-22-5 | |
White | |
99% | |
C8H9NO3·HCl | |
1 g | |
pyridoxal hydrochloride, pyridoxal hcl, vitamin b6 hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride, unii-1416kf0qbc, 2-methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride, pl hcl, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde, hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methyl-4-pyridinecarboxaldehyde hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylpyridine-4-carbaldehyde hydrochloride | |
FCHXJFJNDJXENQ-UHFFFAOYSA-N | |
3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride | |
6171 | |
99% |
Chemical Identifiers
65-22-5 | |
203.62 | |
FCHXJFJNDJXENQ-UHFFFAOYSA-N | |
6171 | |
CC1=NC=C(C(=C1O)C=O)CO.Cl |
C8H9NO3·HCl | |
MFCD00012809 | |
pyridoxal hydrochloride, pyridoxal hcl, vitamin b6 hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride, unii-1416kf0qbc, 2-methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride, pl hcl, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde, hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methyl-4-pyridinecarboxaldehyde hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylpyridine-4-carbaldehyde hydrochloride | |
3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride |
Safety and Handling
EINECSNumber : 200-602-5
RUO – Research Use Only