Please login to your online account to display your discounted pricing

Pyridoxal 5-Phosphate (Monohydrate), Fisher BioReagents


Quantity 5g
Packaging Amber Glass

Manufacturer: fisher scientific company  BP26765

 View more versions of this product

Catalog No. BP26765

  • / Each
  • / Case of 9 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 247.14
Physical Form Powder/Solid
Melting Point 140°C
Quantity 5g
Merck Index 15, 8092
Synonym PLP
Molecular Formula C8H10NO6PH2O
Color Off-white
Assay Percent Range 98%
Packaging Amber Glass
Recommended Storage Store below 0°C
Name Note Monohydrate
CAS 54-47-7
Purity about 98%%
Identification Pass Test
Chemical Name or Material Pyridoxal 5-Phosphate
Infrared Spectrum Conforms to reference
ChemAlert Storage Symbol Gray

Specifications

Chemical Name or Material Pyridoxal 5-Phosphate
Name Note Monohydrate
CAS 54-47-7
Physical Form Powder/Solid
Assay Percent Range 98%
Molecular Formula C8H10NO6PH2O
Formula Weight 247.14

  • PLP is useful for modifying lysyl and valyl residues and functions as a coenzyme in transamination reactions by forming a Schiff-base linkage with lysine groups on aminotransferase
  • PLP is an active form of vitamin B8
  • Also serves as a coenzyme in some decarboxylation and deamination reactions

Accessories
 

Store below 0°C

SDS & Safety Documents