Please login to your online account to display your discounted pricing

Pyridoxal 5-Phosphate (Monohydrate), Fisher BioReagents

$155.32 - $1,372.07

Chemical Identifiers

Chemical Name or Material Pyridoxal 5-Phosphate
Name Note Monohydrate
CAS 54-47-7
Physical Form Powder/Solid
Assay Percent Range 98%
Molecular Formula C8H10NO6PH2O
Formula Weight 247.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BP267625

  SDS
fisher scientific company
BP267625
Amber Glass 25g Each for $1,372.07
Case of 12 Each for $16,464.84

BP26761

  SDS
fisher scientific company
BP26761
Amber Glass 1g Each for $155.32
Case of 9 Each for $1,397.88

BP26765

  SDS
fisher scientific company
BP26765
Amber Glass 5g Each for $399.46
Case of 9 Each for $3,595.14
Description & Specifications

Specifications

Melting Point 140°C
Physical Form Powder/Solid
Formula Weight 247.14
Quantity 25g
Merck Index 15, 8092
Synonym PLP
Molecular Formula C8H10NO6PH2O
Color Off-white
Purity about 98%%
Assay Percent Range 98%
Infrared Spectrum Conforms to reference
ChemAlert Storage Symbol Gray
Identification Pass Test
Packaging Amber Glass
Recommended Storage Store below 0°C
Chemical Name or Material Pyridoxal 5-Phosphate
Name Note Monohydrate
CAS 54-47-7

  • PLP is useful for modifying lysyl and valyl residues and functions as a coenzyme in transamination reactions by forming a Schiff-base linkage with lysine groups on aminotransferase
  • PLP is an active form of vitamin B6
  • Also serves as a coenzyme in some decarboxylation and deamination reactions

Safety and Handling
 

Store below 0°C