missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pyridoxal-5-Phosphate Monohydrate, Approx. 98%, MP Biomedicals™
Manufacturer: Mp Biomedicals 0210276905
Description
CoenzymeSpecifications
Pyridoxal-5-Phosphate Monohydrate | |
5g | |
appox. 98% | |
MFCD00149414 | |
CEEQUQSGVRRXQI-UHFFFAOYSA-N | |
(4-formyl-5-hydroxy-6-methylpyridin-3-yl)methyl dihydrogen phosphate;hydrate | |
38882 | |
Solid |
White | |
41468-25-1 | |
C8H12NO7P | |
4-formyl-5-hydroxy-6-methylpyridin-3-yl methyl dihydrogen phosphate hydrate, phosphopyridoxal monohydrate, pyridoxal 5'-phosphate monohydrate, pyridoxal 5-phosphate monohydrate, pyridoxal phosphate hydrate, pyridoxal-5'-phosphate monohydrate, pyridoxal-5-phosphate monohydrate, pyridoxal-5-phosphate, monohydrate, pyridoxyl phosphate monohydrate, unii-5v5ioj8338 | |
CC1=NC=C(C(=C1O)C=O)COP(=O)(O)O.O | |
265.158 | |
265.1 |
Chemical Identifiers
41468-25-1 | |
265.158 | |
CEEQUQSGVRRXQI-UHFFFAOYSA-N | |
38882 | |
CC1=NC=C(C(=C1O)C=O)COP(=O)(O)O.O |
C8H12NO7P | |
MFCD00149414 | |
4-formyl-5-hydroxy-6-methylpyridin-3-yl methyl dihydrogen phosphate hydrate, phosphopyridoxal monohydrate, pyridoxal 5'-phosphate monohydrate, pyridoxal 5-phosphate monohydrate, pyridoxal phosphate hydrate, pyridoxal-5'-phosphate monohydrate, pyridoxal-5-phosphate monohydrate, pyridoxal-5-phosphate, monohydrate, pyridoxyl phosphate monohydrate, unii-5v5ioj8338 | |
(4-formyl-5-hydroxy-6-methylpyridin-3-yl)methyl dihydrogen phosphate;hydrate |
Safety and Handling
Recommended Storage : Store at -0°C.