missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyridinium Trifluoromethanesulfonate, TCI America™

Manufacturer:  TCI America P162725G

 View more versions of this product

Catalog No. P162725G



Specifications

Specifications

Pyridinium Trifluoromethanesulfonate
224°C
52193-54-1
C6H6F3NO3S
229.17
White
25g
≥98.0% (HPLC,T)
Pyridinium Triflate
Crystal-Powder at 20°C

Chemical Identifiers

52193-54-1
C6H6F3NO3S
Pyridinium Triflate
Safety and Handling

Safety and Handling

TSCA : No

Documents