missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyridinium p-toluenesulfonate, 98+%

Manufacturer:  Alfa Aesar™ A1570814

 View more versions of this product

Catalog No. AAA1570814



Description

Description

Applications
Pyridinium p-Toluenesulfate is used in the synthesis of a subdomain of the cytochrome P450 BM3 enzyme, for use in screening systems for drug candidates. Efficient catalyst for preparing tetrahydropyranyl ethers.

Solubility
Soluble in water (partly), ethanol, acetone, dichloromethane, chloroform, methanol and dimethyl sulfoxide.

Notes
Moisture Sensitive. Store in a tightly closed container. Store in a cool, dry, well-ventilated. Store protected from moisture.
Specifications

Specifications

Pyridinium p-toluenesulfonate
25g
C12H13NO3S
3764305
251.31
116°C to 120°C
24057-28-1
MFCD00013108
pyridinium p-toluenesulfonate, ppts, pyridin-1-ium 4-methylbenzenesulfonate, pyridinium 4-toluenesulfonate, pyridinium tosylate, pyridinium 4-methylbenzenesulfonate, pyridinium p-toluene sulfonate, pyridinium p-toluene sulphonate, pyridinium para-toluenesulfonate, 4-toluenesulfonic acid pyridine salt
≥98%

Chemical Identifiers

24057-28-1
C12H13NO3S
MFCD00013108
pyridinium p-toluenesulfonate, ppts, pyridin-1-ium 4-methylbenzenesulfonate, pyridinium 4-toluenesulfonate, pyridinium tosylate, pyridinium 4-methylbenzenesulfonate, pyridinium p-toluene sulfonate, pyridinium p-toluene sulphonate, pyridinium para-toluenesulfonate, 4-toluenesulfonic acid pyridine salt
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents