Please login to your online account to display your discounted pricing

Pyridinium fluorochromate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Pyridinium fluorochromate
CAS 83042-08-4
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H6CrFNO3
Formula Weight 199.1
MDL Number MFCD00151617
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC302480050

  SDS
thermo fisher scientific
302480050
Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystals
Assay ≥97.5%
Formula Weight 199.1
Color Yellow-orange
Melting Point 156° to 160°C (dec.)
Quantity 5g
Assay Percent Range 98%
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
May cause an allergic skin reaction.
May cause cancer.
May intensify fire; oxidizer.
Chemical Name or Material Pyridinium fluorochromate
Packaging Glass Bottle
Molecular Formula C5H6CrFNO3
MDL Number MFCD00151617
CAS 83042-08-4
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause an allergic skin reaction.
May cause cancer.
May intensify fire; oxidizer.