Learn More
Pyridinium fluorochromate, 98%, Thermo Scientific Chemicals
$52.84 - $142.82
Chemical Identifiers
CAS | 83042-08-4 |
---|---|
Molecular Formula | C5H6CrFNO3 |
MDL Number | MFCD00151617 |
Synonym | pyridin-1-ium; trioxochromium; hydrofluoride, chromium trioxide pyridium hydrofluoride, pyridin-1-ium; tris oxidanylidene chromium; hydrofluoride |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1929506
|
Thermo Scientific Chemicals
L1929506 |
5 g |
Each for $52.84
|
|
|||||
AAL1929514
|
Thermo Scientific Chemicals
L1929514 |
25 g |
Each for $142.82
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
83042-08-4 | |
C5H6CrFNO3 | |
MFCD00151617 | |
pyridin-1-ium; trioxochromium; hydrofluoride, chromium trioxide pyridium hydrofluoride, pyridin-1-ium; tris oxidanylidene chromium; hydrofluoride |
Specifications
83042-08-4 | |
C5H6CrFNO3 | |
5 g | |
Moisture sensitive | |
199.1 | |
Pyridinium fluorochromate |
165°C (decomposition) | |
MFCD00151617 | |
UN1479 | |
pyridin-1-ium; trioxochromium; hydrofluoride, chromium trioxide pyridium hydrofluoride, pyridin-1-ium; tris oxidanylidene chromium; hydrofluoride | |
98% |
Safety and Handling
GHS H Statement
H272-H350-H317
May intensify fire; oxidizer.
May cause cancer.
May cause an allergic skin reaction.
P210-P220-P221-P261-P272-P280-P302+P352-P333+P313-P363-P370+P380+P375-P372-P380-P501c
H272-H317
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
EINECSNumber : 625-358-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only