missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyridinium dichromate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 200882500

 View more versions of this product

Catalog No. AC200882500



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Specifications

Pyridinium dichromate
152.0°C to 153.0°C
Authentic
Plastic bottle
250 g
15,8087
Solubility in water: soluble. Other solubilities: soluble in dmf, dmso and dimethylacetamide -fairly, soluble in acetonenitrile - sparingly soluble in, methylene chloride, chloroform and ether - pract., insol.in hexane,toluene,ethyl ether,ethyl acetate
C1=CC=[NH+]C=C1.C1=CC=[NH+]C=C1.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
376.2
376.2
Crystalline Powder
20039-37-6
Orange to Brown
97.5% min. (Iodometry)
C10H12Cr2N2O7
09,399; 10,335; 12,419; 14,269; 15,57
pyridinium dichromate, cornforth reagent, pyridinium dichromate pdc, oxido-oxido dioxo chromio oxy-dioxochromium; pyridin-1-ium, pyridiniumdichromate, pyridinium-dichromate, dipyridinium dichromate, bis pyridium chromate, pyridiniumdichromate pdc, acmc-209f5g
LMYWWPCAXXPJFF-UHFFFAOYSA-P
oxido-(oxido(dioxo)chromio)oxy-dioxochromium;pyridin-1-ium
2724130
98%

Chemical Identifiers

20039-37-6
376.2
pyridinium dichromate, cornforth reagent, pyridinium dichromate pdc, oxido-oxido dioxo chromio oxy-dioxochromium; pyridin-1-ium, pyridiniumdichromate, pyridinium-dichromate, dipyridinium dichromate, bis pyridium chromate, pyridiniumdichromate pdc, acmc-209f5g
oxido-(oxido(dioxo)chromio)oxy-dioxochromium;pyridin-1-ium
C10H12Cr2N2O7
LMYWWPCAXXPJFF-UHFFFAOYSA-P
2724130
C1=CC=[NH+]C=C1.C1=CC=[NH+]C=C1.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
Safety and Handling

Safety and Handling

GHS H Statement
May intensify fire; oxidizer.
Flammable solid.
May cause cancer by inhalation.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
IF IN EYES: Rins

GHS Signal Word: Danger

EINECSNumber : 243-478-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only