Learn More
Pyridinium dichromate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 200880010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Oxidation reagentSpecifications
Pyridinium dichromate | |
Authentic | |
Plastic bottle | |
1kg | |
15,8087 | |
Solubility in water: soluble. Other solubilities: soluble in dmf, dmso and dimethylacetamide -fairly, soluble in acetonenitrile - sparingly soluble in, methylene chloride, chloroform and ether - pract., insol.in hexane,toluene,ethyl ether,ethyl acetate | |
C1=CC=[NH+]C=C1.C1=CC=[NH+]C=C1.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-] | |
376.2 | |
376.2 |
20039-37-6 | |
97.5% min. (Iodometry) | |
C10H12Cr2N2O7 | |
09,399; 10,335; 12,419; 14,269; 15,57 | |
pyridinium dichromate, cornforth reagent, pyridinium dichromate pdc, oxido-oxido dioxo chromio oxy-dioxochromium; pyridin-1-ium, pyridiniumdichromate, pyridinium-dichromate, dipyridinium dichromate, bis pyridium chromate, pyridiniumdichromate pdc, acmc-209f5g | |
LMYWWPCAXXPJFF-UHFFFAOYSA-P | |
oxido-(oxido(dioxo)chromio)oxy-dioxochromium;pyridin-1-ium | |
2724130 | |
98% |
Safety and Handling
GHS H Statement
May intensify fire; oxidizer.
Flammable solid.
May cause cancer by inhalation.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
IF IN EYES: Rins
GHS Signal Word: Danger
EINECSNumber : 243-478-8
RUO â Research Use Only