missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyridinium bromide perbromide, 90+%, technical, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 158365000

 View more versions of this product

Catalog No. AC158365000



Description

Description

C-X bond formation reagent
Specifications

Specifications

Pyridinium bromide perbromide
Glass bottle
127°C to 133°C
95%
90% min. (Iodometry)
MFCD00013223
01,967; 05,568; 06,499; 09,399; 10,333; 14,268; 15,276
Pyridine hydrobromide, compound with bromine
319.83
Powder, Crystals and/or Chunks
Technical
Authentic
Brown to Orange-Red
500g
39416-48-3
C5H6Br3N
20, V,5, 181
15, 8085
Solubility in water: decomposes. Other solubilities: sparingly soluble in acetic acid
319.83
90+%

Chemical Identifiers

39416-48-3
319.83
Pyridine hydrobromide, compound with bromine
C5H6Br3N
MFCD00013223
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 254-446-8

TSCA : TSCA

SDS Documents