missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyridine, Ultrapure, Spectrophotometric Grade, >99.5%, Alfa Aesar™

Manufacturer:  Alfa Aesar 32436K2

 View more versions of this product

Catalog No. AA32436K2



Specifications

Specifications

Pyridine
Sprectrophotometric
>99.5%
MFCD00011732
JUJWROOIHBZHMG-UHFFFAOYSA-N
pyridine
1049
79.1
0.978g/cm3 at 20°C
20 hPa (15mm Hg) at 20°C
1L
110-86-1
Colorless liquid
C5H5N
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
C1=CC=NC=C1
79.102
CHEBI:16227
Liquid
Unpleasant
Colorless

Chemical Identifiers

110-86-1
79.102
JUJWROOIHBZHMG-UHFFFAOYSA-N
1049
pyridine
C5H5N
MFCD00011732
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
CHEBI:16227
C1=CC=NC=C1
Safety and Handling

Safety and Handling

H225-H302-H312-H332: Highly flammable liquid and vapour. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.

DOTInformation : DOT Class: 3; Packing Group: II

EINECSNumber : 203-809-9

SDS Documents