missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyridine, Ultrapure, Spectrophotometric Grade, >99.5%, Alfa Aesar™

Manufacturer:  Alfa Aesar 32436AK

 View more versions of this product

Catalog No. AA32436AK



Specifications

Specifications

Pyridine
Colorless liquid
C5H5N
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
C1=CC=NC=C1
79.102
CHEBI:16227
Liquid
Unpleasant
Colorless
110-86-1
>99.5%
MFCD00011732
JUJWROOIHBZHMG-UHFFFAOYSA-N
pyridine
1049
79.1
Sprectrophotometric
20 hPa (15mm Hg) at 20°C
250mL

Chemical Identifiers

110-86-1
79.102
JUJWROOIHBZHMG-UHFFFAOYSA-N
1049
pyridine
C5H5N
MFCD00011732
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
CHEBI:16227
C1=CC=NC=C1
Safety and Handling

Safety and Handling

H225-H302-H312-H332: Highly flammable liquid and vapour. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.

DOTInformation : DOT Class: 3; Packing Group: II

EINECSNumber : 203-809-9

SDS Documents