missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pyridine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America P1637100G
Specifications
Pyridine Hydrochloride | |
147°C | |
224°C | |
MFCD00012802 | |
pyridine hydrochloride, pyridine, hydrochloride, pyridinehydrochloride, pyridine hcl, pyridine, hydrochloride 1:1, pyridine-hcl, pyridine.hcl, pyridiniumchloride, pyridine hcl salt, pyrdine hydrochloride | |
C1=CC=NC=C1.Cl | |
115.56 | |
115.56 | |
Crystalline Powder |
628-13-7 | |
White-Yellow | |
C5H6ClN | |
100 g | |
AOJFQRQNPXYVLM-UHFFFAOYSA-N | |
pyridine;hydrochloride | |
10176127 | |
≥98.0% (HPLC,T) |
Chemical Identifiers
628-13-7 | |
115.56 | |
AOJFQRQNPXYVLM-UHFFFAOYSA-N | |
10176127 | |
C1=CC=NC=C1.Cl |
C5H6ClN | |
MFCD00012802 | |
pyridine hydrochloride, pyridine, hydrochloride, pyridinehydrochloride, pyridine hcl, pyridine, hydrochloride 1:1, pyridine-hcl, pyridine.hcl, pyridiniumchloride, pyridine hcl salt, pyrdine hydrochloride | |
pyridine;hydrochloride |
Safety and Handling
RTECSNumber : UT4375000
TSCA : Yes