Learn More
Pyridine hydrobromide, 98%, Thermo Scientific Chemicals
$37.08 - $271.10
Chemical Identifiers
CAS | 18820-82-1 |
---|---|
Molecular Formula | C5H6BrN |
Molecular Weight (g/mol) | 160.014 |
MDL Number | MFCD00012801 |
InChI Key | BBFCIBZLAVOLCF-UHFFFAOYSA-N |
Synonym | pyridine hydrobromide, pyridinium bromide, pyridine, hydrobromide, pyridine, hydrobromide 1:1, pyridine hbr, pyridinehydrobromide, acmc-1bovh, ksc178o2t, pyridine hydrobromide, purum at |
PubChem CID | 87810 |
IUPAC Name | pyridine;hydrobromide |
SMILES | C1=CC=NC=C1.Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1931418
|
Thermo Scientific Chemicals
A1931418 |
50 g |
N/A
|
|
|||||
AAA1931430
|
Thermo Scientific Chemicals
A1931430 |
250 g |
N/A
|
|
|||||
AAA193140B
|
Thermo Scientific Chemicals
A193140B |
1000 g |
N/A
|
|
|||||
Description
Pyridine hydrobromide may be used in chemical synthesis studies. Pyridine hydrobromide is quite useful for brominations of ketones on a micro or semimicro scale.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPyridine hydrobromide may be used in chemical synthesis studies. Pyridine hydrobromide is quite useful for brominations of ketones on a micro or semimicro scale.
Solubility
Soluble in water.
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
Chemical Identifiers
18820-82-1 | |
160.014 | |
BBFCIBZLAVOLCF-UHFFFAOYSA-N | |
87810 | |
C1=CC=NC=C1.Br |
C5H6BrN | |
MFCD00012801 | |
pyridine hydrobromide, pyridinium bromide, pyridine, hydrobromide, pyridine, hydrobromide 1:1, pyridine hbr, pyridinehydrobromide, acmc-1bovh, ksc178o2t, pyridine hydrobromide, purum at | |
pyridine;hydrobromide |
Specifications
18820-82-1 | |
C5H6BrN | |
50 g | |
pyridine hydrobromide, pyridinium bromide, pyridine, hydrobromide, pyridine, hydrobromide 1:1, pyridine hbr, pyridinehydrobromide, acmc-1bovh, ksc178o2t, pyridine hydrobromide, purum at | |
BBFCIBZLAVOLCF-UHFFFAOYSA-N | |
pyridine;hydrobromide | |
87810 | |
98% |
216°C to 220°C | |
MFCD00012801 | |
Hygroscopic | |
Soluble in water. | |
C1=CC=NC=C1.Br | |
160.014 | |
160.01 | |
Pyridine hydrobromide |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 242-600-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only