Please login to your online account to display your discounted pricing

Pyridine-d5, for NMR, with 0.03% TMS, 100.0 atom% D, ACROS Organics™

$317.63 - $317.63

Chemical Identifiers

Chemical Name or Material Pyridine-d5
Name Note 100.0 atom% D
CAS 7291-22-7
CAS 75-76-3
Purity Grade For NMR
Molecular Formula C5D5N
Formula Weight 84.14
MDL Number MFCD00044639
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC351490050

  SDS
thermo fisher scientific
351490050
Glass bottle 5mL Each for $317.63
Description & Specifications

Specifications

Chemical Name or Material Pyridine-d5
Name Note 100.0 atom% D
CAS 7291-22-7
Deuterium (D) 99.95% min.
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Highly flammable liquid and vapor.
CAS 75-76-3
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
EINECS Number 230-720-2
Water 0.02% max. (Coulometric)
Density 1.0500g/mL
Purity Grade For NMR
Physical Form Liquid
Quantity 5mL
Color Clear colorless
Molecular Formula C5D5N
Formula Weight 84.14
MDL Number MFCD00044639
Beilstein 20, III, 2305

For NMR

Safety and Handling

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Highly flammable liquid and vapor.