missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyridine - D5, deuteration degree min 99.8% for NMR spectroscopy MagniSolv™, MilliporeSigma™

Manufacturer:  MilliporeSigma 1074750009

 View more versions of this product

Catalog No. M1074750009



Specifications

Specifications

Pyridine - D5
Characteristic
20 hPa at 20°C (68°F)
Colorless
7291-22-7
C5H5N
JUJWROOIHBZHMG-RALIUCGRSA-N
2,3,4,5,6-pentadeuteriopyridine
558519
Liquid
482°C (900°F)
Glass ampoule
0.97mPa·s (20°C/68°F)
10 x 0.75mL
99.8% min.
pyridine-d5, 2h5 pyridine, c5d5n, 2 h? pyridine, pyridine d5, pyridine-d5, pyridine,crude,light, pyridine, perdeutero, ~2~h_5_ pyridine, de85c
C1=CC=NC=C1
84.133
84.13

Chemical Identifiers

7291-22-7
84.133
pyridine-d5, 2h5 pyridine, c5d5n, 2 h? pyridine, pyridine d5, pyridine-d5, pyridine,crude,light, pyridine, perdeutero, ~2~h_5_ pyridine, de85c
2,3,4,5,6-pentadeuteriopyridine
C5H5N
JUJWROOIHBZHMG-RALIUCGRSA-N
558519
C1=CC=NC=C1
Documents
Promotions

Promotions