Please login to your online account to display your discounted pricing

Pyridine - D5, deuteration degree min 99.8% for NMR spectroscopy MagniSolv™, EMD Millipore™

$249.88 - $302.64

Chemical Identifiers

Chemical Name or Material Pyridine - D5
CAS 7291-22-7
Physical Form Liquid
Assay Percent Range 99.8% min.
Molecular Formula C5D5N
Formula Weight 84.13
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1074750010

 
emd millipore corporation
1074750010
Septum ampoule 10mL Each for $302.64

M1074750009

 
emd millipore corporation
1074750009
Glass ampoule 10 x 0.75mL Each for $249.88
Description & Specifications

Specifications

Chemical Name or Material Pyridine - D5
CAS 7291-22-7
Odor Characteristic
Packaging Septum ampoule
Ignition Point 482°C (900°F)
Vapor Pressure 20 hPa at 20°C (68°F)
Viscosity 0.97mPa&middnot;s (20°C/68°F)
Physical Form Liquid
Quantity 10mL
Assay Percent Range 99.8% min.
Color Colorless
Molecular Formula C5D5N
Formula Weight 84.13